missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Tweezertrode™ Electrode Kit
Tweezertrode™ Electrode Kit features reusable, tweezer-style, in vivo electrodes suitable for drug or gene delivery in animals.
£1340.00 - £1760.00
Specifications
For Use With (Application) | In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery. |
---|---|
Material | Stainless Steel |
Product Line | BTX Electroporation |
Product Type | Electrode Kit |
Voltage | 1000 V |
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Price | Quantity & Availability | |||||
15487280
|
BTX™
450165 |
7 mm |
£1760.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15497280
|
BTX™
450166 |
10 mm |
£1340.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel or platinum at the tip
- Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
- Can be easily cleaned
- Kits are supplied with BTX524 coaxial cable for connection to electroporation generators
Specifications
In vivo applications, including in utero and ex-vivo gene transfection, eletroporation therapy, and transdermal drug delivery. | |
BTX Electroporation | |
1000 V | |
Cable | |
12 mm | |
1 Ea. | |
<0.04 in. to >0.79 in. | |
11.5 cm | |
Banana Cables |
Stainless Steel | |
Electrode Kit | |
Circular | |
0.47 in. | |
10 μsec. to 99 msec. in PBS | |
20 to 80% | |
<1 mm to >2 cm | |
Non-sterile | |
Square Wave |
Intended for Research use only. Not for use in diagnostic or clinical procedures.