missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Drying Flasks for Rotary Evaporators

Special shaped flasks for powder drying

Brand:  BUCHI 000454

Code : A0

Additional Details : Weight : 0.62416kg

 View more versions of this product

Product Code. 12034592

  • £190.00 / Each
Estimated Shipment: 12-08-2024
to see stock.
Add to basket

Description

Description

  • For use with the Rotavapor R 3 and R-210/R-215
  • Available in multiple sizes with different standard joints

Specifications

Specifications

CE
Powder Drying Flask
Plastic and Glass
29.2/32
2000 mL
Standard Glass
Rotary evaporator
29.2/32
60°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers