Learn More
BUCHI Glass Columns C-690
Brand:  BUCHI 044045
 View more versions of this product
Product Code. 12739485
  • £1760.00 / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
49 mm
460 mm
Borosilicate Glass
Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
290 psi
Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
View accessory products