Learn More
BUCHI Glass Columns C-690
Brand:  BUCHI 044049
 View more versions of this product
Product Code. 12769485
  • £2695.00 / Each
Estimated Shipment: 30-12-2024
to see stock.
Explore more special offers
Add to basket
Specifications
70 mm
460 mm
Borosilicate Glass
Preparative Separations Together with The C-600 Sepacore™ Line of Chromatography Separators
218 psi
Column
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products