missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Concentrated Solution pH2.00 (Glycine), NIST Standard for pH measurement, Fisher Chemical™
£164.00 - £536.00
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10080632
|
Fisher Chemical
J/2800C/05 |
1 Ampule |
£536.00
1 ampoule |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10214390
|
Fisher Chemical
J/2800C/90 |
6 Ampules | N/A | ||||||
Description
Specifications
Colorless | |
2 | |
Liquid |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Substances and mixtures corrosive to metals Category 1
Hazard Statement
- H290-May be corrosive to metals.
Precautionary Statement
- P234-Keep only in original container.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title