missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 3.0 (20°C), Citric acid / sodium hydroxide / sodium chloride solution, with fungicide, traceable to SRM from NIST and PTB, Honeywell Fluka™
Citric acid / sodium hydroxide / sodium chloride solution, with fungicide, traceable to SRM from NIST and PTB
Brand: Honeywell-Fluka 31046-1L
29.79 GBP valid until 2024-12-31
Use promo code "21693" to get your promotional price.
This item is not returnable.
View return policy
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Buffer Solution Ph 3.0 (20 °C) | |
1310-73-2, 77-92-9, 7647-14-5, 7732-18-5 | |
Poly bottle |
With fungicide | |
1 L | |
NONH for all modes of transport |
Safety and Handling
Product Identifier
- Buffer solution pH 3.0 (20 °C)