missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 3.0 (20°C), Citric acid / sodium hydroxide / sodium chloride solution, with fungicide, traceable to SRM from NIST and PTB, Honeywell Fluka™

Citric acid / sodium hydroxide / sodium chloride solution, with fungicide, traceable to SRM from NIST and PTB

£43.98 - £230.76

Specifications

CAS 1310-73-2, 77-92-9, 7647-14-5, 7732-18-5
UN Number NONH for all modes of transport
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15663180
View Documents Promotion Details
Honeywell-Fluka
31046-1L
1 L
£43.98
1 L
Please to purchase this item. Need a web account? Register with us today!
15673180
View Documents Promotion Details
Honeywell Fluka
31046-6X1L
6 x 1 L
£230.76
Pack of 6
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Specifications

1310-73-2, 77-92-9, 7647-14-5, 7732-18-5
NONH for all modes of transport
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Buffer solution pH 3.0 (20 °C)

SDS
Documents

Documents

Product Certifications