missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution pH 4, Phthalate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals
£25.50 - £39.50
Specifications
Chemical Name or Material | Buffer solution pH 4 |
---|---|
CAS | 877-24-7, 7732-18-5, 7487-94-7 |
Concentration | 1.0% (Potassium hydrogen phthalate) |
Physical Form | Liquid |
Molecular Formula | C8H5KO4 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10727913
|
Thermo Scientific Acros
383835000 |
500 mL |
£25.50
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10270023
|
Thermo Scientific Acros
383830010 |
1 L |
£39.50
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution pH 4 | |
1.0% (Potassium hydrogen phthalate) | |
C8H5KO4 | |
Solubility in water: miscible. |
877-24-7, 7732-18-5, 7487-94-7 | |
Liquid | |
MFCD00217672 | |
204.22 |
RUO – Research Use Only