missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Buffered Zinc Formalin Histology Fixative
Enhance nuclear detail in immunohistochemistry techniques with Epredia™ Buffered Zinc Formalin.
Brand: Epredia™ 5701ZF
63.75 GBP valid until 2024-12-31
Use promo code "21628" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
Easy-to-pour bottle helps reduce spills.Specifications
4/Cs. | |
Formalin | |
1 Gallon Bottle |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 1A
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity after single exposure Category 1
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H350-May cause cancer.
- H370-Causes damage to organs.
Precautionary Statement
- P362-Take off contaminated clothing.
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands, and anyexposed skin thoroughly after handling.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formaldehyde, methylalcohol, zinc sulphate, sodium acetate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction