missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffers for NucleoSpin™ 96 DNA Plasma
Additional supply of buffer solutions for use in isolation of cell-free, circulating DNA from up to 2 mL blood plasma in high-throughput 96-well format using the NucleoSpin™ 96 DNA kit or core kit.
Brand: Macherey-Nagel™ 740836.250
This item is not returnable.
View return policy
Description
NucleoSpin™ 96 DNA Plasma
- Isolation of cell-free DNA from up to 2 mL blood plasma (i.e., ‘Liquid Biopsies’)
- Highly efficient recovery of cell-free DNA
- High throughput 96-well format solution
- Manual or automated processing
- Optimized protocol for Cell-Free DNA BCT (‘Streck Tubes’)
- Core Kits contain basic content focused on automation platforms
- Typical downstream applications: NGS, qPCR, ddPCR
Specifications
Yellow | |
Guanidine hydrochloride; Ethanol, denatured; Water | |
250 mL | |
Alcoholic | |
Liquid | |
Binding buffer PMB |
NucleoSpin™ 96 DNA Plasma kit or core kit | |
5.0 to 7.0 | |
50-01-1 | |
Room Temperature (18°C to 25°C) | |
Isolation of cell-free DNA from plasma |