missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Buffers for NucleoSpin™ 96 DNA Plasma
Additional supply of buffer solutions for use in isolation of cell-free, circulating DNA from up to 2 mL blood plasma in high-throughput 96-well format using the NucleoSpin™ 96 DNA kit or core kit.
£47.45 - £71.95
Specifications
For Use With (Equipment) | NucleoSpin™ 96 DNA Plasma kit or core kit |
---|---|
CAS | 50-01-1 |
Content And Storage | Room Temperature (18°C to 25°C) |
Form | Liquid |
For Use With (Application) | Isolation of cell-free DNA from plasma |
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Product Type | Quantity | Price | Quantity & Availability | ||||
15819127
|
Macherey-Nagel™
740836.250 |
Binding buffer PMB | 250 mL |
£71.95
250mL |
Estimated Shipment: 28-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15809127
|
Macherey-Nagel™
740835.125 |
Lysis buffer PML | 125 mL |
£47.45
125mL |
Estimated Shipment: 28-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
NucleoSpin™ 96 DNA Plasma
- Isolation of cell-free DNA from up to 2 mL blood plasma (i.e., ‘Liquid Biopsies’)
- Highly efficient recovery of cell-free DNA
- High throughput 96-well format solution
- Manual or automated processing
- Optimized protocol for Cell-Free DNA BCT (‘Streck Tubes’)
- Core Kits contain basic content focused on automation platforms
- Typical downstream applications: NGS, qPCR, ddPCR
Specifications
NucleoSpin™ 96 DNA Plasma kit or core kit | |
Room Temperature (18°C to 25°C) | |
Isolation of cell-free DNA from plasma |
50-01-1 | |
Liquid |