missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffers for NucleoSpin™ RNA Plus

Additional buffer solutions for use with NucleoSpin™ RNA Plus Kits, for efficient and quick RNA isolation from cells and tissues.

Brand:  Macherey-Nagel™ 740907.30

26.26 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 15849127

  • £33.35 / 30mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

NucleoSpin™ RNA Plus Kits

  • gDNA Removal Columns included – no time consuming rDNase digestion
  • Convenient handling – lysate clearing and gDNA removal using the same column (in one step)
  • Novel lysis buffer – exceptionally efficient, with no mercaptoethanol or TCEP necessary
  • Two column silica membrane system (one for DNA removal, one for RNA isolation)
  • Downstream applications: real-time RT-PCR, Northern blotting, primer extension, array technology, RNase protection assays
Specifications

Specifications

Colorless
Ethyleneglycol diacetate
593-84-0
Room Temperature (18°C to 25°C)
Liquid
Binding Solution BS
NucleoSpin RNA Plus kit
30 mL
Odorless
0 to 16% in Water
RNA isolation from cells and tissue
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers