missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Bulk tubes
GREENER_CHOICE

For use with Bead Mill 4 and Bead Mill 24 homogenizers. Also fits most other commercially available Bead Mills.

Brand:  Fisherbrand™ 15535809

237.63 GBP valid until 2024-12-31
Use promo code "21658" to get your promotional price.


 View more versions of this product

Product Code. 15535809

  • £462.00 / 1 pack
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • RNase/DNase free
  • Processing range: 300μL to 1.8mL
  • Beads not included
Specifications

Specifications

Non-reinforced tubes with screw caps and O-rings
White/Opaque
Bead Mill 4 and 24 Homogenizers
Yes
2 mL
Yes
500/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.