Learn More
Butyric acid sodium salt, 98%, Thermo Scientific Chemicals
£20.10 - £95.48
Chemical Identifiers
CAS | 156-54-7 |
---|---|
Molecular Formula | C4H7NaO2 |
Molecular Weight (g/mol) | 110.09 |
MDL Number | MFCD00002816 |
InChI Key | MFBOGIVSZKQAPD-UHFFFAOYSA-M |
Synonym | sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 |
PubChem CID | 5222465 |
ChEBI | CHEBI:64103 |
SMILES | [Na+].CCCC([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10063330
|
Thermo Scientific Acros
263190050 |
5 g | Glass bottle |
£20.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10277110
|
Thermo Scientific Acros
263190250 |
25 g | Glass bottle |
£20.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10474715
|
Thermo Scientific Acros
263191000 |
100 g | Glass bottle |
£28.94
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13458488
|
Thermo Scientific Acros
263195000 |
500 g | Glass Bottle |
£95.48
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
156-54-7 | |
110.09 | |
MFBOGIVSZKQAPD-UHFFFAOYSA-M | |
5222465 | |
[Na+].CCCC([O-])=O |
C4H7NaO2 | |
MFCD00002816 | |
sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 | |
CHEBI:64103 |
Specifications
156-54-7 | |
White | |
C4H7NaO2 | |
MFCD00002816 | |
Solubility in water: soluble. Other solubilities: insoluble in ether | |
[Na+].CCCC([O-])=O | |
110.09 | |
CHEBI:64103 | |
99% | |
Amorphous Powder |
250.0°C to 253.0°C | |
98% | |
CH3CH2CH2COONa | |
sodium butyrate, sodium butanoate, butyric acid sodium salt, butanoic acid, sodium salt, butyrate sodium, butyric acid, sodium salt, sodium n-butyrate, sodium propanecarboxylate, butyric acid, na, butanoic acid, sodium salt 1:1 | |
MFBOGIVSZKQAPD-UHFFFAOYSA-M | |
sodium;butanoate | |
5222465 | |
110.09 | |
Authentic | |
Butyric acid sodium salt |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
- Butyric acid sodium salt
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.