Learn More
Motic™ C-mount Camera Adapter for BA/AE Series Microscope Chip Sensor
C-mount camera adapter compatible with camera having 1/3, 2/3 or 1/2 in. chip sensors. Motic™ C-mount Camera Adapter for BA/AE Series Microscope Chip Sensor fits on the trinocular port of any BA/AE series microscope
Brand:  Motic™ 1101001901791
Code : A0
Additional Details : Weight : 0.09000kg
 View more versions of this product
Product Code. 11803410
  • £237.00 / Each
In Stock
to see stock.
Add to basket
Description
Description
  • Fits the trinocular port of any BA or AE series Microscope:
    • Choose 0.65X magnification with 2/3 in. chip sensor
    • Choose 0.5X magnification with 1/3 or 1/2 in. chip sensor
Specifications
Specifications
C-mount Adapter
8.46, 12.7 mm
For 1/2 in. sensor
0.5X
1/3, 1/2 in.
0.5X
Motic BA210, 310, 410 and AE2000 series compound microscopes
1 Ea.