missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Motic™ C-mount Camera Adapter for BA/AE Series Microscope Chip Sensor
C-mount camera adapter compatible with camera having 1/3, 2/3 or 1/2 in. chip sensors. Motic™ C-mount Camera Adapter for BA/AE Series Microscope Chip Sensor fits on the trinocular port of any BA/AE series microscope
£137.00 - £237.00
Specifications
Type | C-mount Adapter |
---|---|
For Use With (Equipment) | Motic BA210, 310, 410 and AE2000 series compound microscopes |
Quantity | 1 Ea. |
Product Code | Brand | Magnification Power | Diameter (Metric) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Magnification Power | Diameter (Metric) | Price | Quantity & Availability | ||||
11893400
|
Motic™
1101001901781 |
0.65X | 16.9 mm |
£137.00
Each |
Estimated Shipment: 09-08-2024
Log in to see stock. |
||||
11803410
|
Motic™
1101001901791 |
0.5X | 8.46, 12.7 mm |
£237.00
Each |
In Stock
Log in to see stock. |
||||
Description
- Fits the trinocular port of any BA or AE series Microscope:
- Choose 0.65X magnification with 2/3 in. chip sensor
- Choose 0.5X magnification with 1/3 or 1/2 in. chip sensor
Specifications
C-mount Adapter | |
Motic BA210, 310, 410 and AE2000 series compound microscopes | |
1 Ea. |