Learn More
Labcon C-Pette™ and C-Pette™ E Blood Transfer Dispensers
These E dispensers provide an easy way to dispense blood into a test cassette without removing the cap from the collection tube.
Brand:  Labcon 7625-500-000-9
Additional Details : Weight : 0.45360kg
 View more versions of this product
Product Code. 17484362
  • £79.25 / Pack of 500
Estimated Shipment: 05-08-2024
to see stock.
Description
Description
Both E dispensers fit all standard blood collection tubes and are ideal for use with many Point-of-Care (POC) analyzers that require the delivery of a sample.
  • Minimizes aerosols and reduces the risk of lab-acquired infections
  • Safe to use at patient bedside
  • Polystyrene USP Class VI
  • Stainless Steel or Arylic cannula
  • Not Autoclavable
  • Made in USA
Specifications
Specifications
Blood Droplet Dispenser
Yes
Acrylic Cannula
500 Pk.