Learn More
Labcon C-Pette™ and C-Pette™ E Blood Transfer Dispensers
These E dispensers provide an easy way to dispense blood into a test cassette without removing the cap from the collection tube.
£19.45 - £410.00
Product Code | Brand | Quantity | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Includes | Price | Quantity & Availability | ||||
17494362
|
Labcon
7625-540-000-9 |
100 Pk. | Acrylic Cannula |
£19.45
Pack of 100 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17484362
|
Labcon
7625-500-000-9 |
500 Pk. | Acrylic Cannula |
£79.25
Pack of 500 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17424372
|
Labcon
7630-540-000-9 |
100 Pk. | Stainless Steel Cannula |
£115.00
Pack of 100 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17414372
|
Labcon
7630-500-000-9 |
500 Pk. | Stainless Steel Cannula |
£410.00
Pack of 500 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Both E dispensers fit all standard blood collection tubes and are ideal for use with many Point-of-Care (POC) analyzers that require the delivery of a sample.
- Minimizes aerosols and reduces the risk of lab-acquired infections
- Safe to use at patient bedside
- Polystyrene USP Class VI
- Stainless Steel or Arylic cannula
- Not Autoclavable
- Made in USA
Specifications
Blood Droplet Dispenser | |
Yes |