Learn More
Thermo Scientific™ CA(PEG)8 Carboxy-PEG-Amine Compound
CA(PEG)n: pegylated amino acids of the form carboxy-PEG-amine; with 4 to 24 polyethylene glycol units; for surface- and molecule-pegylation applications.
£271.00 - £1026.00
Specifications
Description | Pierce CA(PEG)8 |
---|---|
Content And Storage | Store desiccated at 4°C. Shipped at ambient temperature. |
Label Type | Conjugating Groups, Polyethylene Glycol (PEG) |
Reactive Moiety | Amine, Carboxylic Acid, Primary Amine |
Spacer | 8 ethylene glycol units |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11861415
|
Thermo Scientific™
26122 |
Pierce CA(PEG)8 | 100 mg |
£271.00
100mg |
Estimated Shipment: 26-07-2024
Log in to see stock. |
||||
10177783
|
Thermo Scientific™
26123 |
Pierce CA(PEG)8 | 1 g |
£1026.00
1g |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Thermo Scientific™ Pierce CA(PEG)n Compounds are pegylated amino acids of the form carboxy-PEG-amine, where n equals 4 to 24 polyethylene glycol units, and are useful for a variety of surface- and molecule-pegylation applications.
CA(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with carboxy (-COOH) and amino (-NH2) termini. The unbranched, hydrophilic, discrete-length molecules have the form Carboxy-PEGn-Amine, where the subscript “n” denotes 4, 8, 12, or 24 ethylene glycol units. The carboxyl and primary amine of each compound provide specific targets for crosslinking and other conjugation methods, making these compounds useful as PEGylation reagents.
Highlights:
- Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
- Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Allows site-specific labeling or crosslinking involving primary amines or carboxyl groups on proteins or surfaces
- Easy-to-follow instructions increase the likelihood of a successful outcome
Ordering Information:
Alternative names: Amino-dPEG-acid; Amino-PEG-carboxylic acid; Carboxy-PEO-amine
Compatible with:
Methyl-PEG-Amine Compounds, MA(PEG)n; Other Kinds of PEGylation Reagents
Specifications
Pierce CA(PEG)8 | |
Conjugating Groups, Polyethylene Glycol (PEG) | |
8 ethylene glycol units | |
Amine Compound |
Store desiccated at 4°C. Shipped at ambient temperature. | |
Amine, Carboxylic Acid, Primary Amine | |
Water |
For Research Use Only. Not for use in diagnostic procedures.