missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadence Science™ Disposable Plastic (PTFE) Animal Feeding Needles
For critical applications requiring sterile, flexible, single-use feeding needles
Brand: Cadence Science™
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Nontoxic, nonpyrogenic
- PTFE tubing is flexible and softer than other types of tubing
- Low friction protects delicate organs
- Translucent for easy confirmation of reagent delivery
- Plastic luer hubs; soft silicone tips
- Sterile, packaged in pouches
- Five needles per pouch, 20 pouches per box
Specifications
Soft, Flexible Animal Feeding Needle | |
Yes | |
18G | |
1.5 in. | |
0.078 in. |
PTFE | |
For critical applications requiring sterile, flexible, single use feeding needles | |
Sterile | |
0.38 cm | |
0.02 cm |