Learn More
Cadence Science™ Nonflexible Stainless Steel Animal Feeding Needles
For feeding and dosing without injury to sensitive tissues.
Brand:  Cadence Science™ 7924
 View more versions of this product
Product Code. 16227951
  • £722.00 / Pack of 12
Explore more special offers
Specifications
Animal Feeding Needle
Reusable
13G
3 in.
4 mm
Stainless Steel (304)
Animal feeding and dosing without injury to sensitive tissues, ideal for pharmacologists, toxicologists, pathologists and other biomedical researchers working with laboratory animals
Non-sterile
7.62 cm