missing translation for 'onlineSavingsMsg'
Learn More
Cadmium carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals Product Code.: 11360879

Cadmium carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals

Product Code. 11360879
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11360879

Brand: Thermo Scientific Alfa Aesar 010660.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium carbonate is the raw material for the production of pigments for ceramic and frits and for production of cadmium salt. Used as reagents. It is used for intermediates of manufacturing polyester, glass pigment flux, organic catalysts, plastic plasticizer, stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-78-0
Molecular Formula CCdO3
Molecular Weight (g/mol) 172.42
MDL Number MFCD00010918
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3blyShow MoreShow Less
PubChem CID 10564
IUPAC Name cadmium(2+);carbonate
SMILES [Cd++].[O-]C([O-])=O

Specifications

Melting Point 500°C (decomposition)
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.998% (Metals basis)
Solubility Information Soluble in dilute acids,concentrated solutions of ammonium salts. Insoluble in water.
Formula Weight 172.42
Odor Odorless
Density 4.26 g/cm3
Chemical Name or Material Cadmium carbonate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA29S016 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010660.09,010660.18
Lot NumberP25F020 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010660.09,010660.18
Lot NumberP08H045 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010660.09,010660.18
Lot NumberR28A024 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010660.09,010660.18
Lot NumberM30E078 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010660.09,010660.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.