missing translation for 'onlineSavingsMsg'
Learn More
Cadmium Carbonate, Pure, Thermo Scientific Chemicals Product Code.: 15269716

Cadmium Carbonate, Pure, Thermo Scientific Chemicals

Product Code. 15269716
250 g
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy

Product Code. 15269716

Brand: Thermo Scientific Acros 212322500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point 343°C to 412°C
CAS 513-78-0
Color White
Physical Form Solid
Quantity 250 g
Assay Percent Range 60.0 to 63.0% (Cd)
Molecular Formula CCdO3
Linear Formula CdCO3
MDL Number MFCD00010918
UN Number 2570
Fieser 04,67
Merck Index 15,1621
Synonym cadmium carbonate, mikrokalcit, otavite, kalcit, supermikrokalcit, carbonic acid, cadmium salt 1:1, cadmium monocarbonate, chemcarb cdco3, caswell no. 134a, unii-616zvs3bly
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in diluted acids and in concentrated, solutions of nh4-salts
InChI Key GKDXQAKPHKQZSC-UHFFFAOYSA-L
SMILES [Cd++].[O-]C([O-])=O
IUPAC Name cadmium(2+) carbonate
Molecular Weight (g/mol) 172.42
PubChem CID 10564
Formula Weight 172.41
Grade Pure
Concentration or Composition (by Analyte or Components) >95%
Odor Odorless
Appearance Powder
Chemical Name or Material Cadmium Carbonate, Pure
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0412102 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number212322500,212320010
Lot NumberA0419481 Certificate TypeCertificate of Analysis Date25/11/2022 Catalog Number212322500,212320010
Lot NumberA0416666 Certificate TypeCertificate of Analysis Date15/08/2022 Catalog Number212322500,212320010
Lot NumberA0414161 Certificate TypeCertificate of Analysis Date20/05/2022 Catalog Number212322500,212320010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium carbonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
Hazard Statement
  • H372-Causes damage to organs.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H341-Suspected of causing genetic defects.
  • H361-Suspected of damaging fertility or the unborn child.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium Carbonate, Pure, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.