Learn More
Cadmium chloride, anhydrous, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Cadmium chloride is a precursor material for cadmium sulfide, which is a brilliant yellow pigment known as Cadmium Yellow. It is mainly utilized in dyeing, pigments, photocopying and electroplating of cadmium. It is a mild Lewis acid, and can form tetrachlorocadmium species. It is used in the preparation of organocadmium compounds, such as dialkyl or diaryl cadmium, which are useful for the synthesis of ketones.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Boiling Point | 960°C |
Melting Point | 568°C |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.99% (Metals basis) |
Solubility Information | Highly soluble in water. Soluble in dimethyl sulfoxide. Slightly soluble in alcohol. |
Formula Weight | 183.32 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z30K017 | Certificate of Analysis | 31/12/2024 | 011860.14,011860.22 |
X08K027 | Certificate of Analysis | 17/10/2024 | 011860.14,011860.22 |
Z06J023 | Certificate of Analysis | 01/01/2024 | 011860.14,011860.22 |
M20I051 | Certificate of Analysis | 12/04/2022 | 011860.14,011860.22 |
J13T006 | Certificate of Analysis | 17/11/2021 | 011860.14,011860.22 |
Safety and Handling



- Cadmium chloride, anhydrous
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 1
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P201-Obtain special instructions before use.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- MIXTURE LIST-Contains : Cadmium chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.