missing translation for 'onlineSavingsMsg'
Learn More
Cadmium iodide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11335747

Cadmium iodide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11335747
25 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11335747

Brand: Thermo Scientific Alfa Aesar 043442.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium iodide is used in lithography, photography, electroplating and the manufacturing of phosphors, in engraving process, and was used as an antiseptic and astringent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7790-80-9
Molecular Formula CdI2
Molecular Weight (g/mol) 366.223
MDL Number MFCD00010920
InChI Key OKIIEJOIXGHUKX-UHFFFAOYSA-L
Synonym cadmium iodide, cadmium diiodide, unii-2f2upu4kcw, cadmium 2+ diiodide, 2f2upu4kcw, acmc-20aljc, cadmium 2+ ion diiodide, cadmium iodide trace metals basis 25gShow MoreShow Less
PubChem CID 24635
IUPAC Name cadmium(2+);diiodide
SMILES [Cd+2].[I-].[I-]

Specifications

Boiling Point 787°C
Melting Point 387°C to 389°C
Color Brown
Physical Form Powder
Quantity 25 g
UN Number UN2570
Merck Index 14,1621
Solubility Information It is soluble in ethanol,acetone,ether and ammonia.
Formula Weight 366.21
Percent Purity 99.999%
Grade Puratronic™
Odor Odorless
Sensitivity Hygroscopic; Light sensitive
Density 5.67 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium iodide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN20F023 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number043442.14,043442.22
Lot NumberW23K007 Certificate TypeCertificate of Analysis Date03/10/2024 Catalog Number043442.14,043442.22
Lot NumberW11K011 Certificate TypeCertificate of Analysis Date23/09/2024 Catalog Number043442.14,043442.22
Lot NumberW27J007 Certificate TypeCertificate of Analysis Date05/10/2023 Catalog Number043442.14,043442.22
Lot NumberZ21I031 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number043442.14,043442.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium iodide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.