missing translation for 'onlineSavingsMsg'
Learn More
Cadmium oxide, 98.9%, Thermo Scientific Chemicals Product Code.: 11387538

Cadmium oxide, 98.9%, Thermo Scientific Chemicals

Product Code. 11387538
250 g
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11387538

Brand: Thermo Scientific Alfa Aesar 033235.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cadmium oxide is used as an electroplating chemical and in the manufacture of cadmium electrodes. It is a component of silver alloys, phosphors, semiconductors, glass and ceramic glazes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-19-0
Molecular Formula CdO
Molecular Weight (g/mol) 128.41
MDL Number MFCD00010921
InChI Key CFEAAQFZALKQPA-UHFFFAOYSA-N
Synonym cadmium oxide, cadmium monoxide, cadmium fume, aska-rid, kadmu tlenek polish, caswell no. 136aa, ccris 115, epa pesticide chemical code 236200, dsstox_cid_4715Show MoreShow Less
PubChem CID 14782
SMILES [O--].[Cd++]

Specifications

Melting Point 1430°C
Color Red to Brown
Physical Form Powder
Quantity 250 g
Assay Percent Range 98.9%
Solubility Information Practically insoluble in water. Soluble in dilute acids,slowly soluble in ammonium salts
IUPAC Name cadmium(2+) oxidandiide
Formula Weight 128.4
Odor Odorless
Density 8.15 g/cm3 at 20°C
Chemical Name or Material Cadmium oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG06W024 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number033235.A1,033235.30
Lot NumberX12D052 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number033235.A1,033235.30
Lot NumberR13C058 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number033235.A1,033235.30
Lot NumberF13T019 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number033235.A1,033235.30
Lot NumberP18A042 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number033235.A1,033235.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium oxide, 98.9%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.