Learn More
Cadmium rod, 8mm (0.3in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
£184.92 - £759.78
Chemical Identifiers
CAS | 7440-43-9 |
---|---|
Molecular Formula | Cd |
Molecular Weight (g/mol) | 112.41 |
MDL Number | MFCD00010914 |
InChI Key | BDOSMKKIYDKNTQ-UHFFFAOYSA-N |
Synonym | kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compoundsShow More |
PubChem CID | 23973 |
ChEBI | CHEBI:22977 |
IUPAC Name | cadmium |
SMILES | [Cd] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11304788
|
Thermo Scientific Alfa Aesar
043116.DM |
100 mm |
£184.92
10cm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314788
|
Thermo Scientific Alfa Aesar
043116.HR |
5 x 100 mm |
£759.78
5mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-43-9 | |
112.41 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
23973 | |
cadmium |
Cd | |
MFCD00010914 | |
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compoundsShow More | |
CHEBI:22977 | |
[Cd] |
Specifications
7440-43-9 | |
MFCD00010914 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
cadmium | |
23973 | |
99.999% | |
≈0.43g/mm | |
(metals basis) |
Cd | |
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
[Cd] | |
112.41 | |
CHEBI:22977 | |
Puratronic™ | |
Rod | |
Cadmium rod, 8mm (0.3 in.) dia. |
Safety and Handling
GHS H Statement
H330-H350-H372-H341-H361
Fatal if inhaled.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Suspected of damaging fertility or the unborn child.
P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
H330-H341-H350-H361fd-H372-H500
missing translation for 'einecsNumber' : 231-152-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.