Learn More
Cadmium shot, 1-3mm (0.04-0.1in), Puratronic™, 99.9997% (metals basis), Thermo Scientific Chemicals
£105.07 - £1857.24
Chemical Identifiers
CAS | 7440-43-9 |
---|---|
Molecular Formula | Cd |
Molecular Weight (g/mol) | 112.41 |
MDL Number | MFCD00010914 |
InChI Key | BDOSMKKIYDKNTQ-UHFFFAOYSA-N |
Synonym | kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compoundsShow More |
PubChem CID | 23973 |
ChEBI | CHEBI:22977 |
SMILES | [Cd] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11301129
|
Thermo Scientific Alfa Aesar
011351.18 |
50 g |
£105.07
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11321129
|
Thermo Scientific Alfa Aesar
011351.30 |
250 g |
£458.28
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11311129
|
Thermo Scientific Alfa Aesar
011351.A1 |
1 kg |
£1857.24
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-43-9 | |
112.41 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
23973 | |
[Cd] |
Cd | |
MFCD00010914 | |
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compoundsShow More | |
CHEBI:22977 |
Specifications
Silver-White | |
Silvery-white shot | |
99.9997% (Metals basis) | |
MFCD00010914 | |
BDOSMKKIYDKNTQ-UHFFFAOYSA-N | |
cadmium | |
23973 | |
0.00013 hPa | |
Cadmium |
7440-43-9 | |
8.65g/cm3 | |
Cd | |
kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compounds | |
[Cd] | |
112.41 | |
CHEBI:22977 | |
Shot |
Safety and Handling
P201-P202-P260-P264b-P270-P271-P281-P284-P304+P340-P308+P313-P310-P501c
H330-H341-H350-H361fd-H372-H500
missing translation for 'einecsNumber' : 231-152-8
missing translation for 'rtecsNumber' : EU9800000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Cadmium shot
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
Hazard Statement
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Cadmium
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.