missing translation for 'onlineSavingsMsg'
Learn More
Cadmium shot, 3-6mm (0.1-0.2in), 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11343647

Cadmium shot, 3-6mm (0.1-0.2in), 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11343647
50 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11343647

Brand: Thermo Scientific Alfa Aesar 035483.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-43-9
Molecular Formula Cd
Molecular Weight (g/mol) 112.41
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
Synonym kadmium, compounds, colloidal, cadmium, elemental, kadmium german, unii-00bh33gngh, ccris 112, hsdb 282, 00bh33gngh, cadmium and compoundsShow MoreShow Less
PubChem CID 23973
ChEBI CHEBI:22977
IUPAC Name cadmium
SMILES [Cd]

Specifications

Quantity 50 g
Percent Purity 99.999%
Form Shot
Assay (metals basis)
Chemical Name or Material Cadmium shot, 3mm (0.1 in.)

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS05J032 Certificate TypeCertificate of Analysis Date13/06/2023 Catalog Number035483.30,035483.18
Lot NumberY10H058 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number035662.03,035662.18,035483.30,035483.18
Lot NumberM09E052 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number035662.03,035662.18,035483.30,035483.18
Lot NumberJ16L24 Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number035662.03,035662.18,035483.30,035483.18
Lot NumberJ04Q046A Certificate TypeCertificate of Analysis Date18/11/2021 Catalog Number035662.03,035662.18,035483.30,035483.18
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cadmium shot
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350-May cause cancer.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P201-Obtain special instructions before use.
  • P320-Specific treatment is urgent (see on this label).
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium shot, 3-6mm (0.1-0.2in), 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.