missing translation for 'onlineSavingsMsg'
Learn More
Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals Product Code.: 11370909

Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals

Product Code. 11370909
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11370909

Brand: Thermo Scientific Alfa Aesar 010675.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as an electrolyte in a Weston standard cell, a pigment in fluorescent screens. It is used in the manufacture of cadmium salts of fatty acids, pigments, in electroplating of cadmium in electronic circuits. It act as an intermediate, as an accelerator, in the electrode position of nickel and copper, for stabilizing plastics, in cement formation. It has the ability to absorb hydrochloric acid from waste gases from chemical plants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15244-35-6
Molecular Formula CdO4S
Molecular Weight (g/mol) 208.47
MDL Number MFCD00151655
InChI Key QCUOBSQYDGUHHT-UHFFFAOYSA-L
IUPAC Name cadmium(2+) sulfate
SMILES [Cd++].[O-]S([O-])(=O)=O

Specifications

Melting Point >40°C to H2O
Color White
Physical Form Crystals
Quantity 50 g
Assay Percent Range 99.996% (Metals basis)
Solubility Information Soluble in water. Insoluble in alcohol and ethyl acetate.
Formula Weight 208.46 (Anhydrous)
Refractive Index 1.565
Density 3.08
Chemical Name or Material Cadmium sulfate hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU16J015 Certificate TypeCertificate of Analysis Date06/09/2023 Catalog Number010675.09,010675.18
Lot NumberY18I043 Certificate TypeCertificate of Analysis Date07/12/2022 Catalog Number010675.09,010675.18
Lot NumberC09Q31 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010675.09,010675.18
Lot Number23895 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010675.09,010675.18
Lot Number97443 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number010675.09,010675.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium sulfate hydrate, Puratronic™, 99.996% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.