missing translation for 'onlineSavingsMsg'
Learn More
Cadmium telluride, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11387917

Cadmium telluride, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11387917
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11387917

Brand: Thermo Scientific Alfa Aesar 014367.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Semiconductors, phosphors

Semiconductors, phosphorsCadmium telluride finds applications as the semiconductor material in photovoltaics and infrared optical window. It is used in the production of thin film solar cells. It is doped with chlorine and used as a radiation detector for x-rays, alfa and beta particles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1306-25-8
Molecular Formula CdTe
Molecular Weight (g/mol) 240.01
MDL Number MFCD00015998
InChI Key RPPBZEBXAAZZJH-UHFFFAOYSA-N
Synonym cadmium telluride, irtran 6, cadmium monotelluride, unii-stg188wo13, cadmiumtellurid, irtran-6, cadmium telluride metals basis, cadmium telluride trace metals basisShow MoreShow Less
PubChem CID 91501
IUPAC Name tellanylidenecadmium
SMILES [Cd]=[Te]

Specifications

Melting Point 1041°C
Color Black
Physical Form Polycrystalline Powder
Quantity 25 g
UN Number UN3077
Merck Index 14,1629
Solubility Information Insoluble in water and acids except HNO3 in which it is soluble with decomposition.
Formula Weight 240
Percent Purity 99.999%
Odor Odorless
Density 6.20 g/mL
Assay (metals basis)
Chemical Name or Material Cadmium telluride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX30J020 Certificate TypeCertificate of Analysis Date07/11/2023 Catalog Number014367.14,014367.06
Lot NumberR18J008 Certificate TypeCertificate of Analysis Date31/05/2023 Catalog Number014367.14,014367.06
Lot NumberL03L28 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014367.14,014367.06
Lot NumberC12M11 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014367.14,014367.06
Lot NumberC22X033 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number014367.14,014367.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cadmium telluride, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.