missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadmium wire, 0.64mm (0.025in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
15 m
3 m
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 3 m |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Weight | ≈2.8g/m |
Form | Wire |
Assay | (metals basis) |
Chemical Name or Material | Cadmium wire, 0.64mm (0.025 in.) dia. |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Safety and Handling



Product Identifier
- Cadmium wire
- Danger
- Acute toxicity Category 2
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1B
- Reproductive toxicity Category 2
- Specific target organ toxicity Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H330-Fatal if inhaled.
- H341-Suspected of causing genetic defects.
- H350-May cause cancer.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- H372-Causes damage to organs.
- H410-Very toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P201-Obtain special instructions before use.
- P320-Specific treatment is urgent (see on this label).
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Cadmium wire, 0.64mm (0.025in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction