Learn More
Calcium, 99%, granular, Thermo Scientific Chemicals
£21.81 - £438.00
Chemical Identifiers
CAS | 7440-70-2 |
---|---|
Molecular Formula | Ca |
Molecular Weight (g/mol) | 40.08 |
MDL Number | MFCD00010897 MFCD00085314 |
InChI Key | OYPRJOBELJOOCE-UHFFFAOYSA-N |
Synonym | unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate |
PubChem CID | 5460341 |
ChEBI | CHEBI:29320 |
IUPAC Name | calcium |
SMILES | [Ca] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10579253
|
Thermo Scientific Acros
201180050 |
5 g | Plastic Bottle |
£21.81
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10338580
|
Thermo Scientific Acros
201181000 |
100 g | Plastic Bottle |
£134.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10226450
|
Thermo Scientific Acros
201185000 |
500 g | Plastic Bottle |
£118.59
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10053953
|
Thermo Scientific Acros
201180025 |
2.5 kg | Plastic drum | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-70-2 | |
40.08 | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
5460341 | |
calcium |
Ca | |
MFCD00010897 MFCD00085314 | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelateShow More | |
CHEBI:29320 | |
[Ca] |
Specifications
Gray | |
845.0°C | |
99% | |
Ca | |
05,89; 09,1634; 14,71 | |
unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate | |
[Ca] | |
40.08 | |
CHEBI:29320 | |
99% | |
Calcium, 99% |
7440-70-2 | |
1484.0°C | |
Ca | |
MFCD00010897 MFCD00085314 | |
15, 1648 | |
OYPRJOBELJOOCE-UHFFFAOYSA-N | |
calcium | |
5460341 | |
40.07 | |
Granular |
Safety and Handling
GHS H Statement:
In contact with water releases flammable gases.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas. Protect from moisture.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place. Store in a closed container.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 231-179-5
missing translation for 'tsca' : TSCA
- Calcium
Signal Word
- Danger
Hazard Category
- Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
- H261-In contact with water releases flammable gases.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P231+P232-Handle under inert gas. Protect from moisture.
- P223-Do not allow contact with water.
- P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
- P370+P378-In case of fire: Use carbon dioxide to extinguish.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.