missing translation for 'onlineSavingsMsg'
Learn More
Calcium acetate hydrate, 99%, for analysis, Thermo Scientific Chemicals Product Code.: 12645117

Calcium acetate hydrate, 99%, for analysis, Thermo Scientific Chemicals

Product Code. 12645117
1 kg, Plastic bottle
Click to view available options
Quantity:
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 12645117

Brand: Thermo Scientific Acros 447860010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 114460-21-8
Quantity 1 kg
Assay Percent Range 98.5% min. (on dried substance) (Complexometry)
Molecular Formula C4H6CaO4
Linear Formula (CH3CO2)2Ca
MDL Number MFCD00012448
Synonym Acetic acid, calcium salt hydrate
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol
InChI Key VSGNNIFQASZAOI-UHFFFAOYSA-L
SMILES [Ca++].CC([O-])=O.CC([O-])=O
IUPAC Name calcium diacetate
Molecular Weight (g/mol) 158.17
Formula Weight 158.17
Percent Purity 99%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Calcium acetate hydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468858 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number447860010
Lot NumberA0465066 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number447860010
Lot NumberA0464039 Certificate TypeCertificate of Analysis Date27/06/2024 Catalog Number447860010
Lot NumberA0461618 Certificate TypeCertificate of Analysis Date12/04/2024 Catalog Number447860010
Lot NumberA0460361 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number447860010
Product Identifier
  • Calcium acetate hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium acetate hydrate, 99%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.