missing translation for 'onlineSavingsMsg'
Learn More
Calcium acetate monohydrate, ACS reagent, Thermo Scientific Chemicals Product Code.: 12625027

Calcium acetate monohydrate, ACS reagent, Thermo Scientific Chemicals

Product Code. 12625027
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 12625027

Brand: Thermo Scientific Acros 446351000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5743-26-0
Molecular Formula C4H6CaO4·H2O
Molecular Weight (g/mol) 176.19
InChI Key XQKKWWCELHKGKB-UHFFFAOYSA-L
Synonym calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5puShow MoreShow Less
PubChem CID 82163
ChEBI CHEBI:59199
IUPAC Name calcium;diacetate;hydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.[Ca+2]

Specifications

Melting Point 160.0°C
Quantity 100 g
Linear Formula (CH3CO2)2Ca·H2O
Merck Index 15,1649
Solubility Information Solubility in water: soluble
Formula Weight 176.19
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Calcium acetate monohydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467934 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number446351000,446355000
Lot NumberA0469317 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number446351000,446355000
Lot NumberA0454781 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number446351000,446355000
Lot NumberA0453462 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number446351000,446355000
Lot NumberA0411529 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number446351000,446355000
Product Identifier
  • Calcium acetate monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium acetate monohydrate, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.