missing translation for 'onlineSavingsMsg'
Learn More
Calcium bromide, anhydrous, 99.5%, Thermo Scientific Chemicals Product Code.: 11315059

Calcium bromide, anhydrous, 99.5%, Thermo Scientific Chemicals

Product Code. 11315059
50 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11315059

Brand: Thermo Scientific Alfa Aesar 087893.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Photography, Dehydrating Agent, Manufacturing of Mineral Water

Calcium bromide is a dehydrating agent used in photography, food preservatives, wood preservatives, fire retardants and freezing mixtures. It is used as an oxidizer for mercury emissions control. It is widely used for the preparation of drilling fluids, completion fluids and drilling muds formulations. It also finds application in the manufacture of mineral water.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-41-5
Molecular Formula Br2Ca
Molecular Weight (g/mol) 199.886
MDL Number MFCD00010902
InChI Key WGEFECGEFUFIQW-UHFFFAOYSA-L
Synonym calcium bromide, calcium dibromide, kalziumbromid, brocal, 87cny2eebh, ksc377c0p, calciumbromide, calcium 2+ dibromide, calcium bromide, anhydrous 10gShow MoreShow Less
PubChem CID 5360316
ChEBI CHEBI:31338
IUPAC Name calcium;dibromide
SMILES [Ca+2].[Br-].[Br-]

Specifications

Boiling Point 806°C to 812°C
Melting Point 730°C
Color White
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.5%
Solubility Information Soluble in water,methanol,ethanol and acetone. Insoluble in dioxane,chloroform and ether.
Formula Weight 199.9
Sensitivity Hygroscopic
Appearance White to off-white powder or beads
Density 3.353
Chemical Name or Material Calcium bromide, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP24L050 Certificate TypeCertificate of Analysis Date26/03/2025 Catalog Number087893.09,087893.18,087893.30
Lot NumberR29K039 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number087893.09,087893.18,087893.30
Lot NumberR28K025 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number087893.09,087893.18,087893.30
Lot NumberT03J023 Certificate TypeCertificate of Analysis Date10/05/2024 Catalog Number087893.09,087893.18,087893.30
Lot NumberX03J021 Certificate TypeCertificate of Analysis Date11/10/2023 Catalog Number087893.09,087893.18,087893.30
compliance-icons
Product Identifier
  • Calcium bromide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Calcium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium bromide, anhydrous, 99.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.