Learn More
Calcium carbonate, 99+%, ACS reagent, Thermo Scientific Chemicals
£22.50 - £206.00
Chemical Identifiers
CAS | 471-34-1 |
---|---|
Molecular Formula | CCaO3 |
Molecular Weight (g/mol) | 100.09 |
MDL Number | MFCD00010906 |
InChI Key | VTYYLEPIZMXCLO-UHFFFAOYSA-L |
Synonym | calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama |
PubChem CID | 10112 |
ChEBI | CHEBI:3311 |
SMILES | [Ca++].[O-]C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10606504
|
Thermo Scientific Acros
423511000 |
100 g | Plastic bottle |
£22.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10617581
|
Thermo Scientific Acros
423515000 |
500 g | Plastic bottle |
£61.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10463655
|
Thermo Scientific Acros
423510025 |
2.5 kg | Plastic bottle |
£206.00
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
471-34-1 | |
100.09 | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
10112 | |
[Ca++].[O-]C([O-])=O |
CCaO3 | |
MFCD00010906 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
CHEBI:3311 |
Specifications
825.0°C | |
White | |
99% min. (on dried substance) (Complexometry) | |
CaCO3 | |
01,103; 02,57; 04,67; 05,89 | |
calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama | |
VTYYLEPIZMXCLO-UHFFFAOYSA-L | |
calcium;carbonate | |
10112 | |
100.09 | |
ACS Reagent |
471-34-1 | |
Crystalline Powder | |
CCaO3 | |
MFCD00010906 | |
15, 1659 | |
Solubility in water: insoluble. Other solubilities: soluble in diluted acids | |
[Ca++].[O-]C([O-])=O | |
100.09 | |
CHEBI:3311 | |
≥99% | |
Calcium carbonate, 99+% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
HYGROSCOPIC
missing translation for 'einecsNumber' : 207-439-9
missing translation for 'rtecsNumber' : FF9335000
missing translation for 'tsca' : TSCA
RUO – Research Use Only