missing translation for 'onlineSavingsMsg'
Learn More
Calcium carbonate, 99%, for analysis, precipitated, Thermo Scientific Chemicals Product Code.: 10721111

Calcium carbonate, 99%, for analysis, precipitated, Thermo Scientific Chemicals

Product Code. 10721111
500 g, Plastic bottle
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10721111

Brand: Thermo Scientific Acros 219165000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825.0°C
Color White
Physical Form Powder
Quantity 500 g
Assay Percent Range 98.5% min. (on dried substance) (Complexometry)
Linear Formula CaCO3
Fieser 01,103; 02,57; 04,67; 05,89
Merck Index 15, 1659
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in diluted acids
IUPAC Name calcium;carbonate
Formula Weight 100.09
Percent Purity 99%
Grade Analytical
Loss on Drying 0.5% max. (200°C)
Packaging Plastic bottle
Chemical Name or Material Calcium carbonate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466675 Certificate TypeCertificate of Analysis Date10/10/2024 Catalog Number219165000,219160025,219160010
Lot NumberA0466175 Certificate TypeCertificate of Analysis Date18/09/2024 Catalog Number219165000,219160025,219160010
Lot NumberA0464005 Certificate TypeCertificate of Analysis Date03/07/2024 Catalog Number219165000,219160025,219160010
Lot NumberA0430576 Certificate TypeCertificate of Analysis Date27/11/2023 Catalog Number219165000,219160025,219160010
Lot NumberA0457691 Certificate TypeCertificate of Analysis Date09/11/2023 Catalog Number219165000,219160025,219160010
Product Identifier
  • Calcium carbonate
  • Multiple Manufacturers
    Shop now and save up to 97% on top-quality products for your lab needs.
    Promo Code: 25908

    Expires2025-03-31

    Days

    0

    Hours

    20

    Minutes

    6

    Seconds

    43

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium carbonate, 99%, for analysis, precipitated, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.