missing translation for 'onlineSavingsMsg'
Learn More
Calcium Carbonate, Certified AR for Analysis, Fisher Chemical™ Product Code.: 10508230

Calcium Carbonate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10508230
250 g
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy

Product Code. 10508230

Brand: Fisher Chemical C/1120/50

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 825°C
Color White
pH 8 to 9
Physical Form Solid
Quantity 250 g
Formula Weight 100.09g/mol
Packaging Plastic powder jar
Product Identifier
  • Calcium carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium Carbonate, Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.