missing translation for 'onlineSavingsMsg'
Learn More
Calcium carbonate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11300919

Calcium carbonate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11300919
25 g
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11300919

Brand: Thermo Scientific Alfa Aesar 010679.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It forms the major component of ceramic tile adhesives. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
IUPAC Name calcium;carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 800°C (decomposition)
Color White
Physical Form Powder
Quantity 25 g
Merck Index 14,1657
Solubility Information Very low solubility in water.
Formula Weight 100.09
Percent Purity 99.99%
Grade Puratronic™
Odor Odorless
Refractive Index 1.6583
Density 2.930 g/mL
Assay (metals basis)
Chemical Name or Material Calcium carbonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT22K041 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog Number010679.22,010679.14,010679.36,010679.06
Lot NumberM24K009 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog Number010679.22,010679.14,010679.36,010679.06
Lot NumberX04J013 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number010679.22,010679.14,010679.36,010679.06
Lot NumberW19I008 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number010679.22,010679.14,010679.36,010679.06
Lot NumberW15G042 Certificate TypeCertificate of Analysis Date04/08/2022 Catalog Number010679.22,010679.14,010679.36,010679.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium carbonate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.