missing translation for 'onlineSavingsMsg'
Learn More
Calcium fluoride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals Product Code.: 11378617

Calcium fluoride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11378617
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11378617

Brand: Thermo Scientific Alfa Aesar 035742.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calcium fluoride is the commercially important source for fluoride compounds as well as hydrogen fluoride. It is transparent from ultraviolet to infrared region. It is used in spectroscopy, thermal imaging systems and excimer lasers; it is also used in the manufacturing of optical components including windows, prisms and lenses. In addition, it is also used in aluminum-metallurgy, brake lining manufacturing industries, and in tooth enamel.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-75-5
Molecular Formula CaF2
Molecular Weight (g/mol) 78.08
MDL Number MFCD00010907
InChI Key WUKWITHWXAAZEY-UHFFFAOYSA-L
Synonym calcium fluoride, kalziumfluorid, kalziumdifluorid, calcium 2+ difluoride, acmc-1bk0c, ksc377c5h, calcium fluoride 100g, calcium fluoride,optical gradeShow MoreShow Less
PubChem CID 84512
ChEBI CHEBI:35437
SMILES [F-].[F-].[Ca++]

Specifications

Boiling Point ca 2500°C
Melting Point 1403°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (Metals basis)
Beilstein EM>5,
Solubility Information Slightly soluble in acid. Insoluble in acetone.
IUPAC Name calcium difluoride
Formula Weight 78.08
Odor Odorless
Refractive Index 1.434
Sensitivity Hygroscopic
Density 3.18
Chemical Name or Material Calcium fluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ27K073 Certificate TypeCertificate of Analysis Date31/12/2024 Catalog Number035742.22,035742.14,035742.06
Lot NumberS26F037 Certificate TypeCertificate of Analysis Date25/11/2024 Catalog Number035742.22,035742.14,035742.06
Lot NumberX08K058 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog Number035742.22,035742.14,035742.06
Lot NumberU07K016 Certificate TypeCertificate of Analysis Date20/08/2024 Catalog Number035742.22,035742.14,035742.06
Lot NumberS05K021 Certificate TypeCertificate of Analysis Date25/06/2024 Catalog Number035742.22,035742.14,035742.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium fluoride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.