missing translation for 'onlineSavingsMsg'
Learn More

Calcium granules, -9 mesh, 98.8% (metals basis), Thermo Scientific Chemicals

£31.96 - £269.34

Chemical Identifiers

CAS 7440-70-2
Molecular Formula Ca
Molecular Weight (g/mol) 40.08
MDL Number MFCD00010897 MFCD00085314
InChI Key OYPRJOBELJOOCE-UHFFFAOYSA-N
Synonym unii-sy7q814vup, hsdb 273, sy7q814vup, compounds, elemental, calcium, elemental, kalzium, atom, ingot, chelate
PubChem CID 5460341
ChEBI CHEBI:29320
IUPAC Name calcium
SMILES [Ca]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11318798
SDS View Documents
Thermo Scientific Alfa Aesar
041653.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11328798
SDS View Documents
Thermo Scientific Alfa Aesar
041653.30
250 g
£88.24
250g
Please to purchase this item. Need a web account? Register with us today!
11338798
SDS View Documents
Thermo Scientific Alfa Aesar
041653.A1
1 kg
£269.34
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P223-P231+P232-P280-P335+P334-P370+P378q-P501c

H261

missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: II

missing translation for 'einecsNumber' : 213-179-5

missing translation for 'rtecsNumber' : EV8040000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium granules, -9 mesh, 98.8% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.