Learn More
Calcium hydride, 92% min, Thermo Scientific Chemicals
Description
Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 600°C (decomposition) |
Color | Gray |
Physical Form | Granules |
Quantity | 100 g |
Assay Percent Range | 92% min. |
Solubility Information | Decomposes in water,alcohol and acids to H2Soluble in water and alcohol. Insoluble in benzene. |
Formula Weight | 42.1 |
Sensitivity | Moisture sensitive |
Appearance | Light gray solid |
Density | 1.9 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
X24K010 | Certificate of Analysis | 11/11/2024 | 041590.22,041590.36 |
X20I001 | Certificate of Analysis | 26/10/2022 | 041590.22,041590.36 |
D25M25 | Certificate of Analysis | 08/12/2021 | 041590.22,041590.36 |
X17F009 | Certificate of Analysis | 08/12/2021 | 041590.22,041590.36 |
H12M34 | Certificate of Analysis | 08/12/2021 | 041590.22,041590.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.