Learn More
Calcium hydride, 92% min, Thermo Scientific Chemicals
£132.66 - £314.57
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348718
|
Thermo Scientific Alfa Aesar
041590.22 |
100 g |
£132.66
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358718
|
Thermo Scientific Alfa Aesar
041590.36 |
500 g |
£314.57
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium hydride is used as an efficient drying agent for aprotic base-stable solvents like ethers and tertiary amines. It is a useful dehydrating agent in a synthesis of aldehyde enamines in high yield and purity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7789-78-8 | |
CaH2 | |
MFCD00010897 |
Specifications
600°C (decomposition) | |
Gray | |
92% min. | |
MFCD00010897 | |
42.1 | |
Light gray solid | |
Calcium hydride |
7789-78-8 | |
Granules | |
CaH2 | |
Decomposes in water,alcohol and acids to H2Soluble in water and alcohol. Insoluble in benzene. | |
Moisture sensitive | |
1.9 |
Safety and Handling
P223-P231+P232-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P335+P334-P362-P370+P378q-P501c
H260-H315-H319
missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: I
missing translation for 'einecsNumber' : 232-189-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only