Learn More
Calcium hydride, ca. 93%, extra pure, 0-2 mm grain size, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 675.0°C |
Quantity | 500 g |
Assay Percent Range | 92% min. (Total Ca) |
Linear Formula | CaH2 |
Fieser | 01,105 |
Merck Index | 15,1674 |
Solubility Information | Solubility in water: reacts. Other solubilities: decomposes with lower alcohols and carboxylic acid, soluble only in salt melts |
Formula Weight | 42.09 |
Percent Purity | ∼93% |
Grade | Extra Pure |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0467192 | Certificate of Analysis | 31/01/2025 | 199905000,199900025,199901000 |
A0461915 | Certificate of Analysis | 16/05/2024 | 199905000,199900025,199901000 |
A0458367 | Certificate of Analysis | 13/12/2023 | 199905000,199900025,199901000 |
A0438173 | Certificate of Analysis | 25/08/2023 | 199905000,199900025,199901000 |
A0396681 | Certificate of Analysis | 15/12/2022 | 199905000,199900025,199901000 |
Safety and Handling


- Calcium hydride
- Danger
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Substance which\, in contact with water\, emits flammable gases Category 1
- H260-In contact with water releases flammable gases which may ignite spontaneously.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- EUH014-Reacts violently with water.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Calcium hydride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.