missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calcium oxalate monohydrate, 98%, extra pure, Thermo Scientific Chemicals
£23.32 - £237.18
Chemical Identifiers
CAS | 5794-28-5 |
---|---|
Molecular Formula | C2H2CaO5 |
Molecular Weight (g/mol) | 146.11 |
MDL Number | MFCD00150039 |
InChI Key | LQHWSGSWNOHVHO-UHFFFAOYSA-L |
Synonym | calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:? |
PubChem CID | 165350 |
IUPAC Name | calcium;oxalate;hydrate |
SMILES | O.[Ca++].[O-]C(=O)C([O-])=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10537742
|
Thermo Scientific Acros
403880050 |
5 g | Glass bottle |
£23.32
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10442771
|
Thermo Scientific Acros
403885000 |
500 g | Glass bottle |
£73.57
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10197243
|
Thermo Scientific Acros
403880025 |
2.5 kg | Plastic bottle |
£237.18
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5794-28-5 | |
146.11 | |
LQHWSGSWNOHVHO-UHFFFAOYSA-L | |
165350 | |
O.[Ca++].[O-]C(=O)C([O-])=O |
C2H2CaO5 | |
MFCD00150039 | |
calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:? | |
calcium;oxalate;hydrate |
Specifications
200.0°C | |
White | |
98% | |
(COO)2Ca·H2O | |
2811 | |
calcium oxalate monohydrate, unii-4pp86kk527, calcium hydrate oxalate, calcium ethanedioate hydrate 1:1:1, acmc-1bmv0, ethanedioic acid, calcium salt 1:1 , monohydrate, calcium oxalate monohydrate 100g, ethanedioic acid,calcium salt, hydrate 1:?:? | |
LQHWSGSWNOHVHO-UHFFFAOYSA-L | |
calcium;oxalate;hydrate | |
165350 | |
98% | |
13% max. (200°C) |
5794-28-5 | |
Powder | |
C2H2CaO5 | |
MFCD00150039 | |
14, 1685 | |
Solubility in water: practically insoluble. Other solubilities: practically insoluble in acetic acid,soluble in dilute hcl or hno3 | |
O.[Ca++].[O-]C(=O)C([O-])=O | |
146.11 | |
146.11 | |
Extra Pure | |
Calcium oxalate monohydrate, 98% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Calcium oxalate monohydrate
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
RUO – Research Use Only