missing translation for 'onlineSavingsMsg'
Learn More
Calcium oxide, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11400041

Calcium oxide, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11400041
1 kg
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11400041

Brand: Thermo Scientific Alfa Aesar 044776.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Calcium oxide has wide-ranging utility in industry. It is used in making porcelain, cements, mortars and glass. It is also employed in flue-gas desulfurization in power plants, purifying sugar, drying the solvents/reagents, preparing bleaching powder, calcium carbide and calcium cyanamide. It also finds use in water softeners, chemical recovery at Kraft pulp mills, biodiesel production and in the petroleum industry for the detection of traces of water. In agriculture, it is used to restore the pH of acidic soils (liming). It is useful as a basic catalyst and an acid scavenger in several organic transformations, for example, in Arndt-Eistert synthesis of alpha-diazoketones, synthesis of heterocycles and oximes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1305-78-8
Molecular Formula CaO
Molecular Weight (g/mol) 56.08
MDL Number MFCD00010911
InChI Key BRPQOXSCLDDYGP-UHFFFAOYSA-N
Synonym calcium oxide, burnt lime, calcia, calxyl, gebrannter kalk, unslaked lime, calcium monoxide, airlock, calx
PubChem CID 14778
ChEBI CHEBI:31344
SMILES [O--].[Ca++]

Specifications

Boiling Point 2850°C
Melting Point 2572°C
Color White
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.95% (Metal basis)
Solubility Information Soluble in water. Insoluble in methanol,diethyl ether and n-octanol.
IUPAC Name oxocalcium
Formula Weight 56.08
Refractive Index 1.83
Sensitivity Air and moisture sensitive
Appearance White powder
Density 3.37
Chemical Name or Material Calcium oxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX29H054 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number044776.30,044776.A1,044776.18
Lot NumberE26X038 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044776.30,044776.A1,044776.18
Lot NumberZ01C026 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044776.30,044776.A1,044776.18
Lot NumberG27X016 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044776.30,044776.A1,044776.18
Lot NumberH23R070 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number044776.30,044776.A1,044776.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Calcium oxide, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.