Learn More
Calcium peroxide, typically 65%, Thermo Scientific Chemicals
Description
Calcium peroxide is used as a rubber stabilizer, used in different areas of industry and agriculture. In agriculture it is used as an oxygen fertilizer, and is also used in the presowing treatments of rice seed. It is used as flour bleaching agent and improving agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Physical Form | Powder |
Quantity | 100 g |
UN Number | UN1457 |
Merck Index | 14,1689 |
Solubility Information | Slightly soluble in water; Soluble in acids |
Formula Weight | 72.08 |
Percent Purity | 65% (typically) |
Odor | Odorless |
Sensitivity | Moisture sensitive |
Chemical Name or Material | Calcium peroxide |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T06J039 | Certificate of Analysis | 13/07/2023 | 021157,021157.22,021157.36 |
M20J012 | Certificate of Analysis | 26/01/2023 | 021157.22,021157.36 |
X03I063 | Certificate of Analysis | 13/10/2022 | 021157.22,021157.36 |
D15X016 | Certificate of Analysis | 07/12/2021 | 021157.22,021157.36 |
R21H002 | Certificate of Analysis | 07/12/2021 | 021157.22,021157.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.