Learn More
Calcium peroxide, typically 65%, Thermo Scientific Chemicals
Rubber stabilizer
£45.53 - £121.61
Chemical Identifiers
CAS | 1305-79-9 |
---|---|
Molecular Formula | CaO2 |
Molecular Weight (g/mol) | 72.076 |
MDL Number | MFCD00015988 |
InChI Key | LHJQIRIGXXHNLA-UHFFFAOYSA-N |
Synonym | calcium peroxide, calcium dioxide, calper, calper g, unii-7fro2eno91, peroxidocalcium, calciumperoxid, kalziumperoxid, calcium peroxide ca o2, hsdb 965Show More |
PubChem CID | 14779 |
ChEBI | CHEBI:48233 |
IUPAC Name | calcium;peroxide |
SMILES | [O-][O-].[Ca+2] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11348007
|
Thermo Scientific Alfa Aesar
021157.22 |
100 g |
£45.53
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11358007
|
Thermo Scientific Alfa Aesar
021157.36 |
500 g |
£121.61
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Calcium peroxide is used as a rubber stabilizer, used in different areas of industry and agriculture. In agriculture it is used as an oxygen fertilizer, and is also used in the presowing treatments of rice seed. It is used as flour bleaching agent and improving agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1305-79-9 | |
72.076 | |
LHJQIRIGXXHNLA-UHFFFAOYSA-N | |
14779 | |
calcium;peroxide |
CaO2 | |
MFCD00015988 | |
calcium peroxide, calcium dioxide, calper, calper g, unii-7fro2eno91, peroxidocalcium, calciumperoxid, kalziumperoxid, calcium peroxide ca o2, hsdb 965Show More | |
CHEBI:48233 | |
[O-][O-].[Ca+2] |
Specifications
1305-79-9 | |
CaO2 | |
UN1457 | |
calcium peroxide, calcium dioxide, calper, calper g, unii-7fro2eno91, peroxidocalcium, calciumperoxid, kalziumperoxid, calcium peroxide ca o2, hsdb 965 | |
LHJQIRIGXXHNLA-UHFFFAOYSA-N | |
calcium;peroxide | |
14779 | |
72.08 | |
Odorless | |
Calcium peroxide |
Powder | |
MFCD00015988 | |
14,1689 | |
Slightly soluble in water; Soluble in acids | |
[O-][O-].[Ca+2] | |
72.076 | |
CHEBI:48233 | |
65% (typically) | |
Moisture sensitive |
Safety and Handling
GHS H Statement
H272-H314-H318
May intensify fire
oxidizer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P220-P221-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H272-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: CALCIUM PEROXIDE
missing translation for 'einecsNumber' : 215-139-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.