Learn More
Calcium sulfate, anhydrous, 99%, Thermo Scientific Chemicals
£24.92 - £301.50
Chemical Identifiers
CAS | 7778-18-9 |
---|---|
Molecular Formula | CaO4S |
Molecular Weight (g/mol) | 136.13 |
MDL Number | MFCD00010912 |
InChI Key | OSGAYBCDTDRGGQ-UHFFFAOYSA-L |
Synonym | calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrousShow More |
PubChem CID | 24497 |
ChEBI | CHEBI:31346 |
IUPAC Name | calcium;sulfate |
SMILES | [Ca++].[O-]S([O-])(=O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11379287
|
Thermo Scientific Alfa Aesar
040144.30 |
250 g |
£24.92
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11389287
|
Thermo Scientific Alfa Aesar
040144.A1 |
1 kg |
£61.91
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11369287
|
Thermo Scientific Alfa Aesar
040144.A7 |
5 kg |
£301.50
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as Portland-cement retarder; tile and plaster; It is the source of sulfur and sulfuric acid; Also used in polishing powder and paints, dyeing and calico printing. It is likewise employed in zinc metallurgy. It is employed in drying industrial gases, solids and many organic liquids. In granulated form it is used as soil conditioner. Used for food additives in the soy manufacturing industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7778-18-9 | |
136.13 | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
24497 | |
calcium;sulfate |
CaO4S | |
MFCD00010912 | |
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrousShow More | |
CHEBI:31346 | |
[Ca++].[O-]S([O-])(=O)=O |
Specifications
1450°C | |
White | |
99% | |
MFCD00010912 | |
Soluble in water,slightly soluble in glycerol. | |
[Ca++].[O-]S([O-])(=O)=O | |
136.13 | |
CHEBI:31346 | |
Hygroscopic | |
2.96 g/cm3 at 20°C |
7778-18-9 | |
Powder | |
CaO4S | |
calcium sulfate, drierite, gypsum, sulfuric acid, calcium salt 1:1, karstenite, calcium sulphate, anhydrous gypsum, plaster of paris, anhydrous calcium sulfate, calcium sulfate, anhydrous | |
OSGAYBCDTDRGGQ-UHFFFAOYSA-L | |
calcium;sulfate | |
24497 | |
136.14 | |
White powder | |
Calcium sulfate, anhydrous |
Safety and Handling
H315-H319 : Causes skin irritation. Causes serious eye irritation.
missing translation for 'einecsNumber' : 231-900-3
missing translation for 'rtecsNumber' : WS6920000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.